Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCBP2 (hnRNP E2), Mouse anti-Human, Clone: PC 81-1, Millipore Sigma™

Catalog No. MABF212225 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF212225 25 μg
MABF2122100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF212225 Supplier MilliporeSigma Supplier No. MABF212225UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PCBP2 (hnRNP E2), clone PC81-1, Cat. No. MABF2122, is a mouse monoclonal antibody that detects Poly(rC)-binding protein 2 and has been tested for use in Immunocytochemistry and Western Blotting.

Poly(rC)-binding protein 2 (UniProt: Q15366; also known as Alpha-CP2, Heterogeneous nuclear ribonucleoprotein E2, hnRNP E2) is encoded by the PCBP2 gene (Gene ID: 5094) in human. PCBP2 belongs to a class of cellular proteins that bind to poly(C) stretches of both RNA and DNA and has been detected in all tissues studied. PCBP2 shuttles between the nucleus and cytoplasm and exerts its impact on RNA processing and mRNA expression through sequence-specific interactions with C-rich determinants within target mRNAs. It acts as an important mediator of multiple processes, including replication of viruses with gastrointestinal and hepatic tropism, hepatic collagen synthesis, globin expression, and cellular proliferation. PCBP2 is shown to negatively regulate cellular antiviral responses mediated by mitochondrial antiviral-signaling protein (MAVS) signaling and acts as an adapter between MAVS and the E3 ubiquitin ligase ITCH, thereby triggering MAVS ubiquitination and degradation. Together with PCBP1, it is shown to function as a translational coactivator of poliovirus RNA via a sequence-specific interaction with stem-loop IV of the IRES, promoting poliovirus RNA replication. It has also been implicated in translational control of the 15-lipoxygenase mRNA, human papillomavirus type 16 L2 mRNA, and hepatitis A virus RNA. PCBP2 contains three K-homologous (KH) domains that mediates poly(C) binding. Eight different isoforms of PCBP2 have been described that are produced by alternative splicing. (Ref.: Ghanem, LR et al. (2014). Gene Exp. Patterns 14(2); 78-87; Sean, P., et al. (2008). Virology. Virology. 378(2); 243-253).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCBP2 (hnRNP E2)
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone PC 81-1
Conjugate Unconjugated
Dilution Western Blotting Analysis: A representative lot detected PCBP2 (hnRNP E2) in Western Blotting applications (Sean, P., et. al. (2008). Virology. 378(2):243-53; Chase, A.J., et. al. (2014). Virology. 449:35-44).Immunocytochemistry Analysis: A representative lot detected PCBP2 (hnRNP E2) in Immunocytochemistry applications (Fitzgerald, K.D., et. at. (2011). PLoS Pathog. 7(7):e1002127).Western Blotting Analysis: 1μg/mL from a representative lot detected PCBP2 (hnRNP E2) in SK-N-SH cell lysate.
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Poly(rC)-binding protein 2;Alpha-CP2;Heterogeneous nuclear ribonucleoprotein E2;hnRNP E2
Gene Symbols PCBP2
Host Species Mouse
Immunogen His-tagged full-length human recombinant Poly(rC)-binding protein 2.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001122383
Test Specificity Clone PC 81-1 specifically detects Poly(rC)-binding protein 2 in human cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.