Learn More
Invitrogen™ PCBP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113710
Description
Immunogen sequence: QTPFPPLGQT NPAFPGEKLP LHSSEEAQNL MGQSSGLDAS PPASTHELTI For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
This gene encodes a member of the KH-domain protein subfamily. Proteins of this subfamily, also referred to as alpha-CPs, bind to RNA with a specificity for C-rich pyrimidine regions. Alpha-CPs play important roles in post-transcriptional activities and have different cellular distributions. This gene's protein is found in the cytoplasm, yet it lacks the nuclear localization signals found in other subfamily members. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications
PCBP3 | |
Polyclonal | |
Unconjugated | |
PCBP3 | |
alpha-CP3; AlphaCP-3; Pcbp3; PCBP3OT; PCBP3-OT1; PCBP3-overlapping transcript; PCBP3-overlapping transcript 1; poly(rC) binding protein 3; poly(rC)-binding protein 3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
54039 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P57721 | |
PCBP3 | |
Synthetic peptide directed towards the middle region of human PCBP3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.