Learn More
Invitrogen™ PCBP4 Polyclonal Antibody

Description
The antibody detects endogenous levels of total MCG10 protein.
Specifications
Specifications
| Antigen | PCBP4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | PCBP4 |
| Gene Accession No. | P57723, P57724 |
| Gene Alias | 1200003L19Rik; alpha CP-4; alpha-CP4; AlphaCP-4; alphaCP-4 protein; CBP; LIP4; LYST-interacting protein; MCG10; Pcbp4; poly(rC) binding protein 4; poly(rC)-binding protein 4; RNA binding protein MCG10 |
| Gene Symbols | PCBP4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.