Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCBP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566104
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566104 Supplier Invitrogen™ Supplier No. PA566104
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATIPYHPSLSL GTVLLSANQG FSVQGQYGAV TPAEVTKLQQ LSSHAVPFAT PSVVPGLDPG TQTSSQEFLV P Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.

MCG10 is a member of the KH-domain RNA-binding protein family. Proteins of this family also referred to as alpha-CPs, bind to RNA with specificity for C-rich pyrimidine regions. Alpha-CPs play important roles in post-transcriptional activities and have different cellular distributions. MCG10 is induced by p53 and DNA damage via two p53-responsive promoter elements. Its overexpression in transfected cells results in the suppression of cell proliferation by inducing apoptosis and cell cycle arrest in G2-M. Decreased expression of one isoform of MCG10 has been observed in many forms of lung cancer, leading to speculation that MCG10 can act as a tumor suppressor. Multiple alternatively spliced isoforms of MCG10 have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCBP4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCBP4
Gene Accession No. P57723
Gene Alias 1200003L19Rik; alpha CP-4; alpha-CP4; AlphaCP-4; alphaCP-4 protein; CBP; LIP4; LYST-interacting protein; MCG10; Pcbp4; poly(rC) binding protein 4; poly(rC)-binding protein 4; RNA binding protein MCG10
Gene Symbols PCBP4
Host Species Rabbit
Immunogen Recombinant Human PCBP4. Recombinant protein control fragment (Product #RP-105083).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57060
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.