Learn More
PCCA Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304324AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
This protein is a key enzyme in the catabolic pathway of odd-chain fatty acids, isoleucine, threonine, methionine, and valine. It is probably a dodecamer composed of six biotin-containing alpha subunits and six beta subunits. The protein is located in the mitochondrial matrix. Defects in PCCA are the cause o fpropionic acidemia type I (PA-1) [MIM:606054]. PA-1 is a life-threatening disease characterized by episodic vomiting, lethargy and ketosis, neutropenia, periodic thrombocytopenia, hypogammaglobulinemia, develop-mental retardation, and intolerance to protein.Specifications
PCCA | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P05165, Q91ZA3 | |
PCCA | |
Between 678 and 728 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PCCA | |
RP11-151A6.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
110821, 5095 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.