Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCCB Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1A9A5, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 665011IG150UL

Description
The protein encoded by this gene is a subunit of the propionyl-CoA carboxylase (PCC) enzyme, which is involved in the catabolism of propionyl-CoA. PCC is a mitochondrial enzyme that probably acts as a dodecamer of six alpha subunits and six beta subunits. This gene encodes the beta subunit of PCC. Defects in this gene are a cause of propionic acidemia type II (PA-2).Specifications
PCCB | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P05166, P07633, P79384, Q99MN9 | |
PCCB | |
PCCB Fusion Protein Ag1608 | |
150 μL | |
Primary | |
Human, Mouse, Pig, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1A9A5 | |
Unconjugated | |
PCCB | |
PCCase subunit beta, PCCB | |
Mouse | |
Protein G | |
RUO | |
100158147, 24624, 5096, 66904 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction