Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584011
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584011 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584011 Supplier Invitrogen™ Supplier No. PA584011
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TRVVYKVPEE QPPNTLIGSL AADYGFPDVG HLYKLEVGAP YLRVDGKTGD IFTTETSIDR EGLRECQNQL PGDPCILEFE VSITDLVQN Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 94%.

Protocadherin 1, also known as PCDH1 and cadherin-like protein 1, is a transmembrane protein involved in cell-cell adhesion. Protocadherin 1 belongs to a group of cadherin-like proteins termed delta1-protocadherins, which consists of protocadherin-1, -7, -9, and -11(X/Y). The delta1-protocadherins have seven extracellular cadherin-repeats, a transmembrane domain, and a cytoplasmic domain, and can be expressed in two isoforms, short or long. Expression of protocadherin 1 is regulated both temporally and spatially, and plays a role both in early development and mature tissues. In mice, it is expressed in a number of tissues and organs, including the brain, spinal cord, peripheral ganglia, inner ear, hair follicles, kidney, reproductive organs, prostate, and the seminal gland. Notably, protocadherin 1 is a susceptibility gene for Bronchial Hyperresponsiveness in humans. Expression of protocadherin 1 has also been identified on human pluripotent stem cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDH1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCDH1
Gene Accession No. Q08174
Gene Alias 2010005A06Rik; AI585920; cadherin-like 1; cadherin-like protein 1; PC42; PCDH1; PCDH42; protocadherin 1; protocadherin 1 (cadherin-like 1); protocadherin 42; protocadherin-1; Protocadherin-42
Gene Symbols PCDH1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PCDH1. Recombinant protein control fragment (Product #RP-98501).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5097
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.