Learn More
PCDH1 (Protocadherin 1) Monoclonal Antibody (CSTEM31), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$1562.00
Specifications
Antigen | PCDH1 (Protocadherin 1) |
---|---|
Clone | CSTEM31 |
Concentration | 0.5 mg/mL |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
14-915-737
![]() |
Invitrogen™
14915737 |
2 mg |
Each of 1 for $1,562.00
|
|
|||||
Description
This monoclonal antibody CSTEM31 reacts with human Protocadherin-1, also known as PCDH1 and cadherin-like protein 1. Applications Reported: This CSTEM31 antibody has been reported for use in immunocytochemical staining and flow cytometric analysis. (Fluorochrome-conjugated CSTEM31 is recommended for use in flow cytometery.) Applications Tested: This CSTEM31 antibody has been tested by immunocytochemistry on formaldehyde-fixed and permeabilized human iPSCs, and by flow cytometry on human iPSCs. For immunocytochemistry, this can be used at less than or equal to 5 μg/mL. For flow cytometry, this antibody can be used at less than or equal to 0.5 μg/test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Protocadherin 1, also known as PCDH1 and cadherin-like protein 1, is a transmembrane protein involved in cell-cell adhesion. Protocadherin 1 belongs to a group of cadherin-like proteins termed delta1-protocadherins, which consists of protocadherin-1, -7, -9, and -11(X/Y). The delta1-protocadherins have seven extracellular cadherin-repeats, a transmembrane domain, and a cytoplasmic domain, and can be expressed in two isoforms, short or long. Expression of protocadherin 1 is regulated both temporally and spatially, and plays a role both in early development and mature tissues. In mice, it is expressed in a number of tissues and organs, including the brain, spinal cord, peripheral ganglia, inner ear, hair follicles, kidney, reproductive organs, prostate, and the seminal gland. Notably, protocadherin 1 is a susceptibility gene for Bronchial Hyperresponsiveness in humans. Expression of protocadherin 1 has also been identified on human pluripotent stem cells.Specifications
PCDH1 (Protocadherin 1) | |
0.5 mg/mL | |
Unconjugated | |
Mouse | |
Human | |
Q08174 | |
5097 | |
Primary | |
4°C, do not freeze | |
PCDH1 |
CSTEM31 | |
Monoclonal | |
Liquid | |
RUO | |
PBS with proprietary stabilizer and 0.09% sodium azide; pH 7.2 | |
2010005A06Rik; AI585920; cadherin-like 1; cadherin-like protein 1; PC42; PCDH1; PCDH42; protocadherin 1; protocadherin 1 (cadherin-like 1); protocadherin 42; protocadherin-1; Protocadherin-42 | |
IgG2b κ | |
Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.