Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDH10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567292
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567292 Supplier Invitrogen™ Supplier No. PA567292
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPSFVPSDGRQ AADYRSNLHV PGMDSVPDTE VFETPEAQPG AERSFSTFGK EKALHSTLER KELDGLLTNT Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The mRNA encodes a cadherin-related neuronal receptor thought to play a role in the establishment and function of specific cell-cell connections in the brain. This family member contains 6 extracellular cadherin domains, a transmembrane domain and a cytoplasmic tail differing from those of the classical cadherins. Alternatively spliced transcripts encode isoforms with unique cytoplasmic domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDH10
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCDH10
Gene Accession No. Q9P2E7
Gene Alias 6430521D13Rik; 6430703F07Rik; KIAA1400; LOW QUALITY PROTEIN: protocadherin-10; mKIAA1400; OL-pc; OL-PCDH; OL-protocadherin; ortholog of OL-pcdh; PCDH10; PCDH19; protocadherin 10; protocadherin-10; RGD1565811
Gene Symbols PCDH10
Host Species Rabbit
Immunogen Recombinant Human PCDH10. Recombinant protein control fragment (Product #RP-107931).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57575
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.