Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCDH11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP180939
Description
PCDH11 Polyclonal specifically detects PCDH11 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PCDH11 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
PCDH11, PCDH11Y, PCDH22, PCDH-PC, PCDHX, PCDH-X, PCDHY, PCDH-Y, protein phosphatase 1, regulatory subunit 119, protocadherin 11X, protocadherin 22, Protocadherin on the X chromosome, Protocadherin on the Y chromosome, Protocadherin prostate cancer, protocadherin-11 X-linked, Protocadherin-11 Y-linked, Protocadherin-22, protocadherin-PC, Protocadherin-S | |
Rabbit | |
Affinity Purified | |
RUO | |
27328 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
PCDH11 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:QFKLVYKTGDVPLIRIEEDTGEIFTTGARIDREKLCAGIPRDEHCFYEVEVAILPDEIFRLVKIRFLIEDINDNAPLFPATVINISIPENSAINSKYTLPAAVDPDVGINGVQNYELIKSQNIFGLDVIETPEG | |
0.1 mL | |
Primary | |
The immunogen sequence has 100% identity to both PCDH11X and PCDH11Y. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction