Learn More
Invitrogen™ PCDH11X/PCDH11Y Polyclonal Antibody

Description
Additional Information: This antibody is expected to recognize all reported Y-linked isoforms (NP_116753.1; NP_116754.1; NP_116755.1) and all reported X-linked isoforms (NP_055337.1; NP_055337.1; NP_116750.1; NP_116751.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
Specifications
Specifications
| Antigen | PCDH11X/PCDH11Y |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | PCDH11X |
| Gene Accession No. | Q9BZA7, Q9BZA8 |
| Gene Alias | KIAA1326; PCDH11; PCDH11X; PCDH11Y; PCDH22; PCDH-PC; PCDHX; PCDH-X; PCDHY; PCDH-Y; PPP1R119; protein phosphatase 1, regulatory subunit 119; protocadherin 11 X-linked; protocadherin 11 Y-linked; protocadherin 11X; protocadherin 22; protocadherin on the X chromosome; Protocadherin on the Y chromosome; protocadherin prostate cancer; Protocadherin-11; Protocadherin-11 X-linked; protocadherin-11 Y-linked; Protocadherin-22; Protocadherin-PC; protocadherin-S |
| Gene Symbols | PCDH11X, PCDH11Y |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.