Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDH11X Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981108
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1108 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1108 Supplier Bioss Supplier No. BS11107R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein consists of an extracellular domain containing 7 cadherin repeats, a transmembrane domain and a cytoplasmic tail that differs from those of the classical cadherins. The gene is located in a major X/Y block of homology and its Y homolog, despite divergence leading to coding region changes, is the most closely related cadherin family member. The protein is thought to play a fundamental role in cell-cell recognition essential for the segmental development and function of the central nervous system. Disruption of this gene may be associated with developmental dyslexia. Alternative splicing results in multiple transcript variants.

Specifications

Antigen PCDH11X
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PCDH11X
Gene Accession No. Q9BZA7
Gene Alias KIAA1326; PCDH11; PCDH11X; PCDH11Y; PCDHX; PCDH-X; PCDHX11; PPP1R119; protein phosphatase 1, regulatory subunit 119; protocadherin 11 X-linked; protocadherin 11 Y-linked; protocadherin 11X; Protocadherin on the X chromosome; protocadherin X-linked 11; Protocadherin-11; Protocadherin-11 X-linked; Protocadherin-S; RGD1562864
Gene Symbols PCDH11X
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PCDH11X.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27328
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.