Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDH11Y Monoclonal Antibody (1G5)
GREENER_CHOICE

Catalog No. PIMA522690
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522690 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522690 Supplier Invitrogen™ Supplier No. MA522690
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: EKNYTIREEI PENVLIGNLL KDLNLSLIPN KSLTTTMQFK LVYKTGDVPL IRIEEDTGEI FTTGARIDRE KLCAGIPRDE HCFYEVEVAI LPDEIFRLVK IRFLIEDIN.

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein consists of an extracellular domain containing 7 cadherin repeats, a transmembrane domain and a cytoplasmic tail that differs from those of the classical cadherins. The gene is located in a major X/Y block of homology and its most closely related cadherin superfamily member is located in this homologous region on the X chromosome. The protein is thought to play a fundamental role in cell-cell recognition essential for the segmental development and function of the central nervous system. Transcripts arising from alternative splicing encode isoforms with N- and C-terminal variation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDH11Y
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1G5
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PCDH11Y
Gene Accession No. Q9BZA8
Gene Alias KIAA1326; PCDH11; PCDH11X; PCDH11Y; PCDH22; PCDH-PC; PCDHX; PCDH-X; PCDHY; PCDH-Y; PPP1R119; protein phosphatase 1, regulatory subunit 119; protocadherin 11 X-linked; protocadherin 11 Y-linked; protocadherin 11X; protocadherin 22; protocadherin on the X chromosome; Protocadherin on the Y chromosome; protocadherin prostate cancer; Protocadherin-11; Protocadherin-11 X-linked; protocadherin-11 Y-linked; Protocadherin-22; Protocadherin-PC; protocadherin-S
Gene Symbols PCDH11Y
Host Species Mouse
Immunogen PCDH11Y (NP_004733, 57 a.a. ~ 165 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83259
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.