Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDH11X/PCDH11Y Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5103355
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103355 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103355 Supplier Thermo Scientific Supplier No. PA5103355

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PCDH-X/Y.

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein consists of an extracellular domain containing 7 cadherin repeats, a transmembrane domain and a cytoplasmic tail that differs from those of the classical cadherins. The gene is located in a major X/Y block of homology and its most closely related cadherin superfamily member is located in this homologous region on the X chromosome. The protein is thought to play a fundamental role in cell-cell recognition essential for the segmental development and function of the central nervous system. Transcripts arising from alternative splicing encode isoforms with N- and C-terminal variation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDH11X/PCDH11Y
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PCDH11X
Gene Accession No. Q9BZA7, Q9BZA8
Gene Alias KIAA1326; PCDH11; PCDH11X; PCDH11Y; PCDH22; PCDH-PC; PCDHX; PCDH-X; PCDHY; PCDH-Y; PPP1R119; protein phosphatase 1, regulatory subunit 119; protocadherin 11 X-linked; protocadherin 11 Y-linked; protocadherin 11X; protocadherin 22; protocadherin on the X chromosome; Protocadherin on the Y chromosome; protocadherin prostate cancer; Protocadherin-11; Protocadherin-11 X-linked; protocadherin-11 Y-linked; Protocadherin-22; Protocadherin-PC; protocadherin-S
Gene Symbols PCDH11X, PCDH11Y
Host Species Rabbit
Immunogen A synthesized peptide derived from human PCDH11Y(Accession Q9BZA8), corresponding to amino acid residues L1304-L1340.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27328, 83259
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.