Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCDH11Y Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PCDH11Y |
---|---|
Concentration | 0.18 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2680
![]() ![]() |
Proteintech Group Inc
200691AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody can recognize PCDH11X and PCDH11Y.
This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein consists of an extracellular domain containing 7 cadherin repeats, a transmembrane domain and a cytoplasmic tail that differs from those of the classical cadherins. The gene is located in a major X/Y block of homology and its most closely related cadherin superfamily member is located in this homologous region on the X chromosome. The protein is thought to play a fundamental role in cell-cell recognition essential for the segmental development and function of the central nervous system. Transcripts arising from alternative splicing encode isoforms with N- and C-terminal variation.Specifications
PCDH11Y | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9BZA7, Q9BZA8 | |
27328, 83259 | |
Peptide | |
Primary | |
-20°C | |
PCDH11Y |
0.18 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
PCDH PC, PCDH Y, PCDH11, PCDH11X, PCDH11Y, PCDH22, PCDHY, Protocadherin 11, protocadherin 11 Y linked, Protocadherin 22, Protocadherin PC, Protocadherin prostate cancer | |
PCDH11X, PCDH11Y | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title