Learn More
PCDH15 Mouse anti-Human, Clone: OTI7F10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF812881

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
PCDH15 is a member of the cadherin superfamily. Family members encode integral membrane proteins that mediate calcium-dependent cell-cell adhesion. PCDH15 consists of a signal peptide, 11 extracellular calcium-binding domains, a transmembrane domain and a unique cytoplasmic domain. It plays an essential role in maintece of normal retinal and cochlear function. Mutations in this gene have been associated with hearing loss, which is consistent with its location at the Usher syndrome type 1F (USH1F) critical region on chromosome 10.Specifications
PCDH15 | |
Monoclonal | |
Unconjugated | |
PCDH15 | |
CDHR15, DFNB23, USH1F | |
Mouse | |
Affinity Chromatography | |
RUO | |
65217 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI7F10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96QU1 | |
PCDH15 | |
Human recombit protein fragment corresponding to amino acids 160-400 of human PCDH15 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.