Learn More
Invitrogen™ PCDH15 Polyclonal Antibody

Description
PCDH15 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | PCDH15 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | PCDH15 |
| Gene Accession No. | Q99PJ1 |
| Gene Alias | Ames waltzer; av; BB078305; cadherin-related family member 15; CDHR15; DFNB23; DKFZp667A1711; ENSMUSG00000046980; Gm9815; nmf19; Pcdh15; protocadherin 15; protocadherin 15 CD2; protocadherin 15 CD3 isoform; protocadherin related 15; protocadherin-15; protocadherin-15-CD2; protocadherin-related 15; RP11-449J3.2; USH1F |
| Gene Symbols | PCDH15 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.