Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDH15 Polyclonal Antibody

Catalog No. PIPA5121704
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121704 20 μL
PIPA5119378 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121704 Supplier Invitrogen™ Supplier No. PA5121704
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PCDH15 Polyclonal Antibody for Western Blot, IHC (P)

PCDH15 is a member of the cadherin superfamily. Family members encode integral membrane proteins that mediate calcium-dependent cell-cell adhesion. PCDH15 consists of a signal peptide, 11 extracellular calcium-binding domains, a transmembrane domain and a unique cytoplasmic domain. It plays an essential role in maintenance of normal retinal and cochlear function. Mutations in this gene have been associated with hearing loss, which is consistent with its location at the Usher syndrome type 1F (USH1F) critical region on chromosome 10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDH15
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PCDH15
Gene Accession No. Q99PJ1
Gene Alias Ames waltzer; av; BB078305; cadherin-related family member 15; CDHR15; DFNB23; DKFZp667A1711; ENSMUSG00000046980; Gm9815; nmf19; Pcdh15; protocadherin 15; protocadherin 15 CD2; protocadherin 15 CD3 isoform; protocadherin related 15; protocadherin-15; protocadherin-15-CD2; protocadherin-related 15; RP11-449J3.2; USH1F
Gene Symbols PCDH15
Host Species Rabbit
Immunogen Recombinant protein Protocadherin 15. The antigen corresponds to amino acid range 45-270 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11994
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.