Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDH15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116505
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116505 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116505 Supplier Invitrogen™ Supplier No. PA5116505
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: SH-SY5Y, 293T, A-549, Raji, Mouse brain, Mouse liver, Mouse spleen, Rat brain, Rat liver Immunogen sequence: TVNELTPVGT TIFTGFSGDN GATDIDDGPN GQIEYVIQYN PDDPTSNDTF EIPLMLTGNI VLRKRLNYED KTRYFVIIQA NDRAQNLNER RTTTTTLTVD VLDGDDLGPM FLPCVLVPNT RDCRPLTYQA AIPELRTPEE LNPIIVTPPI QAIDQDRNIQ PPSDRPGILY SILVGTPEDY PRFFHMHPRT AELSLLEPVN RDFHQKFDLV IKAEQDNGHP LPAFAGLHIE ILDENNQSPY F.

PCDH15 is a member of the cadherin superfamily. Family members encode integral membrane proteins that mediate calcium-dependent cell-cell adhesion. PCDH15 consists of a signal peptide, 11 extracellular calcium-binding domains, a transmembrane domain and a unique cytoplasmic domain. It plays an essential role in maintenance of normal retinal and cochlear function. Mutations in this gene have been associated with hearing loss, which is consistent with its location at the Usher syndrome type 1F (USH1F) critical region on chromosome 10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDH15
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PCDH15
Gene Accession No. Q96QU1, Q99PJ1
Gene Alias Ames waltzer; av; BB078305; cadherin-related family member 15; CDHR15; DFNB23; DKFZp667A1711; ENSMUSG00000046980; Gm9815; nmf19; Pcdh15; protocadherin 15; protocadherin 15 CD2; protocadherin 15 CD3 isoform; protocadherin related 15; protocadherin-15; protocadherin-15-CD2; protocadherin-related 15; RP11-449J3.2; USH1F
Gene Symbols PCDH15
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 160-400 of human PCDH15 (NP_0011362351).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11994, 65217, 690865
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.