Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCDH17 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537941
Description
This antibody is predicted to react with human, rat, pig and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein contains six extracellular cadherin domains, a transmembrane domain, and a cytoplasmic tail differing from those of the classical cadherins. The encoded protein may play a role in the establishment and function of specific cell-cell connections in the brain.
Specifications
| PCDH17 | |
| Polyclonal | |
| Unconjugated | |
| PCDH17 | |
| C030033F14Rik; Gm78; PCDH17; PCDH68; PCH68; protocadherin 17; protocadherin 68; protocadherin-17; Protocadherin-68 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 219228, 27253 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O14917 | |
| PCDH17 | |
| Peptide with sequence C-RVFADVHSRASRDS, from the internal region near the C-terminus of PCDH17 (aa1098-1111). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction