Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDH17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110077
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110077 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110077 Supplier Invitrogen™ Supplier No. PA5110077
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRALDSDFGE SGRLTYEIVD GNDDHLFEID PSSGEIRTLH PFWEDVTPVV ELVVKVTDHG KPTLSAVAKL IIRSVSGSLP EGVPRVNGEQ HHWDMSLPLI V.

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein contains six extracellular cadherin domains, a transmembrane domain, and a cytoplasmic tail differing from those of the classical cadherins. The encoded protein may play a role in the establishment and function of specific cell-cell connections in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDH17
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PCDH17
Gene Accession No. O14917
Gene Alias C030033F14Rik; Gm78; PCDH17; PCDH68; PCH68; protocadherin 17; protocadherin 68; protocadherin-17; Protocadherin-68
Gene Symbols PCDH17
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 610-710 of human PCDH17 (NP_001035519.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27253
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.