Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDH18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553686
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553686 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553686 Supplier Invitrogen™ Supplier No. PA553686
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IHKGDITLVP TINGTLPIRS HHRSSPSSSP TLERGQMGSR QSHNSHQSLN SLVTISSNHV PENFSLELTH ATPAVEQVSQ LLSMLHQGQY QPRPSFRGNK YSRSYR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

Protocadherins comprise the largest group within the cadherin family of calcium-dependent cell-cell adhesion molecules. Protocadherin 18 (PCDH18) was initially identified through screening of human and mouse ESTs using conserved cytoplasmic domain motifs. Tissue screening revealed PCDH18 broad expression in tissues such as brain, liver, heart, kidney, lung, and trachea. PCDH18 expression is also temporally and spatially regulated in the mouse embryonic brain and interacts with Disabled-1, an intracellular adapter protein involved in neuronal migration and cell positioning during mammalian brain development. PCDH18 was also found to be expressed in the developing zebrafish neural tube and central nervous system, lending support to the hypothesis that PCDH18 may play a role during brain development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDH18
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCDH18
Gene Accession No. Q9HCL0
Gene Alias 3110038E07Rik; BB095589; DKFZP434B0923; Kiaa1562; PCDH18; PCDH68L; Protocadherin 18; protocadherin 68-like protein; protocadherin-18
Gene Symbols PCDH18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PCDH18. Recombinant protein control fragment (Product #RP-90578).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54510
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.