Learn More
PCDH19 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304468A

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is a member of the delta-2 protocadherin subclass of the cadherin superfamily. The encoded protein is thought to be a calcium-dependent cell-adhesion protein that is primarily expressed in the brain. Mutations in this gene on human chromosome X are associated with sporadic infantile epileptic encephalopathy and to a female-restricted form of epilepsy (EFMR; also known as PCDH19RE). Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
PCDH19 | |
Polyclonal | |
Unconjugated | |
PCDH19 | |
EFMR, EIEE9, epilepsy, female restricted, with mental retardation (Juberg-Hellman syndrome), KIAA1313 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
279653, 57526 | |
4° C | |
Liquid |
Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q80TF3, Q8TAB3 | |
PCDH19 | |
Between 1098 and 1148 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.