Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDH19 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501567799
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7799 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7799 Supplier Bethyl Laboratories, Inc Supplier No. A304468AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The protein encoded by this gene is a member of the delta-2 protocadherin subclass of the cadherin superfamily. The encoded protein is thought to be a calcium-dependent cell-adhesion protein that is primarily expressed in the brain. Mutations in this gene on human chromosome X are associated with sporadic infantile epileptic encephalopathy and to a female-restricted form of epilepsy (EFMR; also known as PCDH19RE). Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen PCDH19
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene PCDH19
Gene Accession No. Q80TF3, Q8TAB3
Gene Alias EFMR, EIEE9, epilepsy, female restricted, with mental retardation (Juberg-Hellman syndrome), KIAA1313
Gene Symbols Pcdh19
Host Species Rabbit
Immunogen Between 1098 and 1148
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 279653, 57526
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.