Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDH20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584560
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584560 Supplier Invitrogen™ Supplier No. PA584560
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GAEWNPPLSF SLASRGLSGQ YVTLDNRSGE LHTSAQEIDR EALCVEGGGG TAWSGSVSIS SSPSDSCLLL LDVLVLPQEY FRFV Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 86%.

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. This gene encodes a protein which contains 6 extracellular cadherin domains, a transmembrane domain and a cytoplasmic tail differing from those of the classical cadherins. Although its specific function is undetermined, the cadherin-related neuronal receptor is thought to play a role in the establishment and function of specific cell-cell connections in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDH20
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCDH20
Gene Accession No. Q8N6Y1
Gene Alias C630015B17Rik; PCDH13; PCDH20; protocadherin 13; protocadherin 20; protocadherin-13; Protocadherin-20
Gene Symbols PCDH20
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PCDH20. Recombinant protein control fragment (Product #RP-108475).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64881
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.