Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCDH7 Antibody (OTI1F7), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273269B
Description
PCDH7 Monoclonal specifically detects PCDH7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
PCDH7 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
PCDH7 | |
Full length recombinant protein of human PCDH7 (NP_115833) produced in HEK293T cells. | |
0.1 mL | |
5099.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1F7 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
BH-PcdhBHPCDHprotocadherin-7, BH-protocadherin (brain-heart), Brain-heart protocadherin, protocadherin 7 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction