Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDH7 Monoclonal Antibody (OTI2G6)
GREENER_CHOICE

Catalog No. PIMA526137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526137 Supplier Invitrogen™ Supplier No. MA526137
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PCDH7 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The gene encodes a protein with an extracellular domain containing 7 cadherin repeats. The gene product is an integral membrane protein that is thought to function in cell-cell recognition and adhesion. Alternative splicing yields isoforms with unique cytoplasmic tails.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDH7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI2G6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PCDH7
Gene Accession No. O60245
Gene Alias BHPCDH; BH-Pcdh; BH-protocadherin (brain-heart); Brain-heart protocadherin; cell adhesion molecule; LOW QUALITY PROTEIN: protocadherin-7; PCDH7; PPP1R120; protein phosphatase 1, regulatory subunit 120; protocadherin 42-like protein; protocadherin 7; protocadherin 7; BH-protocadherin; BH-protocadherin (brain-heart); protocadherin-7; protocadherin-7; LOW QUALITY PROTEIN: protocadherin-7
Gene Symbols PCDH7
Host Species Mouse
Immunogen Full length human recombinant protein of PCDH7 produced in HKE293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5099, 54216
Target Species Human, Mouse, Non-human Primate, Non-human Primate
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.