Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDH7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552862 Supplier Invitrogen™ Supplier No. PA552862
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGEVTFSLES GSEYLKIDNL TGELSTSERR IDREKLPQCQ MIFDENECFL DFEVSVIGPS QSWVDLFEGQ VIVLDINDNT PTFPSPVLTL TVEENRPVGT LYLLPT Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The gene encodes a protein with an extracellular domain containing 7 cadherin repeats. The gene product is an integral membrane protein that is thought to function in cell-cell recognition and adhesion. Alternative splicing yields isoforms with unique cytoplasmic tails.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDH7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCDH7
Gene Accession No. O60245
Gene Alias BHPCDH; BH-Pcdh; BH-protocadherin (brain-heart); Brain-heart protocadherin; cell adhesion molecule; LOW QUALITY PROTEIN: protocadherin-7; PCDH7; PPP1R120; protein phosphatase 1, regulatory subunit 120; protocadherin 42-like protein; protocadherin 7; protocadherin 7; BH-protocadherin; BH-protocadherin (brain-heart); protocadherin-7; protocadherin-7; LOW QUALITY PROTEIN: protocadherin-7
Gene Symbols PCDH7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PCDH7. Recombinant protein control fragment (Product #RP-89864).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5099
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.