Learn More
Invitrogen™ PCDH8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552697
Description
Immunogen sequence: GDSDFNDSDS DISGDALKKD LINHMQSGLW ACTAECKILG HSDRCWSPSC SGPNAHPSPH PPAQMSTFCK STSLPRDPLR RDNYYQAQLP KTVGLQSVYE KVLHRDYDRT VTLLSPPRPG RLPDLQEIGV PLYQSPPGRY LSPKKGANEN Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Specifications
PCDH8 | |
Polyclonal | |
Unconjugated | |
Pcdh8 | |
1700080P15Rik; Activity-regulated cadherin-like protein; arcadlin; PAPC; Paraxial protocadherin; PCDH8; protocadherin 8; protocadherin-8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5100 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95206 | |
Pcdh8 | |
Recombinant protein corresponding to Human PCDH8. Recombinant protein control fragment (Product #RP-90241). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.