Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDH8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589703
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589703 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589703 Supplier Invitrogen™ Supplier No. PA589703
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTAASLDRER IAEYNLTLVA EDRGAPPLRT VRPYTVRVGD ENDNAPLFTR PVYEVSVREN NPPGAYLATV AARDRDLGRN GQVTYRLLEA EVGRAGGAVS TYVSVDPATG AIYALRSFDY ETLRQLDVRI QASDGGSPQL SSSALVQVRV LDQNDHAPVL VHPAPANGSL EVAVPGRTAK DTVVARVQAR DADEGANGEL AFELQQQEPR EAFAIGRRTG EILLTGDLSQ EPPGRVFRAL LVISDGGRPP LTTTATVSFV VTAGGGRGPA APASAGSPER SRPPGSRLGV SGSVLQWDTP L; Positive Samples: U-251MG, SH-SY5Y, Mouse brain, Rat brain; Cellular Location: Cell junction, Cell membrane, Cell projection, Single-pass type I membrane protein, dendrite, postsynaptic cell membrane, presynaptic cell membrane, synapse.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDH8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Pcdh8
Gene Accession No. D3ZE55, O95206, Q7TSK3
Gene Alias 1700080P15Rik; Activity-regulated cadherin-like protein; arcadlin; PAPC; Paraxial protocadherin; PCDH8; protocadherin 8; protocadherin-8
Gene Symbols Pcdh8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 450-750 of human PCDH8 (NP_0025812).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18530, 5100, 64865
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.