Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDH8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547739
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547739 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547739 Supplier Invitrogen™ Supplier No. PA547739
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, less than 2% cross-reactivity with recombinant human Protocadherin-10 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDH8
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Pcdh8
Gene Accession No. O95206
Gene Alias 1700080P15Rik; Activity-regulated cadherin-like protein; arcadlin; PAPC; Paraxial protocadherin; PCDH8; protocadherin 8; protocadherin-8
Gene Symbols Pcdh8
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Protocadherin-8 Lys30-Pro749.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5100
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.