Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCDH9 Antibody (7G3F7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261771
Description
PCDH9 Monoclonal specifically detects PCDH9 in Human samples. It is validated for Western Blot, ELISA.Specifications
PCDH9 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
protocadherin 9, protocadherin-9 | |
Mouse | |
136 kDa | |
0.1 mL | |
5101 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA | |
7G3F7 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
PCDH9 | |
Purified recombinant fragment of human PCDH9 (AA: 24-148) expressed in E. Coli. | |
Protein G purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction