Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDH9 Monoclonal Antibody (7G3A2)
GREENER_CHOICE

Catalog No. PIMA531831
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531831 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531831 Supplier Invitrogen™ Supplier No. MA531831
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31831 has been tested in indirect ELISA.

This gene encodes a member of the protocadherin family, and cadherin superfamily, of transmembrane proteins containing cadherin domains. These proteins mediate cell adhesion in neural tissues in the presence of calcium. The encoded protein may be involved in signaling at neuronal synaptic junctions. Sharing a characteristic with other protocadherin genes, this gene has a notably large exon that encodes multiple cadherin domains and a transmembrane region. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDH9
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7G3A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PCDH9
Gene Accession No. Q9HC56
Gene Alias cadherin superfamily protein VR4-11; Pcdh9; protocadherin 9; protocadherin-9
Gene Symbols PCDH9
Host Species Mouse
Immunogen Purified recombinant fragment of human PCDH9 (AA: 24-148) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5101
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.