Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDH9 Mouse anti-Human, Clone:7G3F7, Abnova™

Catalog No. 89333993 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-993 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-993 Supplier Abnova Corporation Supplier No. MAB17527
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human PCDH9.

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The mRNA encodes a cadherin-related neuronal receptor that localizes to synaptic junctions and is putatively involved in specific neuronal connections and signal transduction. Sharing a characteristic with other protocadherin genes, this gene has a notably large exon that encodes six cadherin domains and a transmembrane region. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen PCDH9
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7G3F7
Conjugate Unconjugated
Description protocadherin 9
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry
Flow Cytometry
Immunohistochemistry
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene PCDH9
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 24-148 of human PCDH9 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 5101
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.