Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDH9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120292
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120292 Supplier Invitrogen™ Supplier No. PA5120292
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: A-549, U-251MG, SH-SY5Y, NCI-H460. The target is usually found in the following locations: Cell membrane, Single-pass type I membrane protein. Immunogen sequence: ICFIEREVPF HLKAVYDNQY LLETSSLLDY EGTKEFSFKI VASDSGKPSL NQTALVRVKL EDENDNPPIF NQPVIELSVS ENNRRGLYLT TISATDEDSG KNADIVYQLG PNASFFDLDR KTGVLTASRV FDREEQERFI FTVTARDNGT PPLQSQAAVI VTVLDENDNS PKFTHNHFQF FVSENLPKYS TVGVITVTDA D.

This gene encodes a member of the protocadherin family, and cadherin superfamily, of transmembrane proteins containing cadherin domains. These proteins mediate cell adhesion in neural tissues in the presence of calcium. The encoded protein may be involved in signaling at neuronal synaptic junctions. Sharing a characteristic with other protocadherin genes, this gene has a notably large exon that encodes multiple cadherin domains and a transmembrane region. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDH9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PCDH9
Gene Accession No. Q9HC56
Gene Alias cadherin superfamily protein VR4-11; Pcdh9; protocadherin 9; protocadherin-9
Gene Symbols PCDH9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 400-600 of human PCDH9 (NP_982354.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5101
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.