Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCDHA11 Polyclonal Antibody
PCDHA11 Polyclonal Antibody
Supplier: Thermo Scientific PA569597
Description
PCDHA11 Polyclonal Antibody for Western Blot

Specifications
PCDHA11 | |
Polyclonal | |
Unconjugated | |
PCDHA11 | |
A830022B16Rik; cadherin-related neuronal receptor 11; cadherin-related neuronal receptor 7; CNR7; CNRN7; CNRS7; CRNR7; KIAA0345-like 3; LOW QUALITY PROTEIN: protocadherin alpha-10; PCDHA11; PCDH-ALPHA11; PCDH-alpha-11; PCDH-alpha9 homolog; protocadherin alpha 11; protocadherin alpha 9 homolog; protocadherin alpha-11 | |
Rabbit | |
Affinity chromatography | |
RUO | |
56138 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y5I1 | |
PCDHA11 | |
synthetic peptide directed towards the middle region of human PCDHA11 (aa 171-220). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction