Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDHA11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567445
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567445 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567445 Supplier Invitrogen™ Supplier No. PA567445
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VNGEVTYSLMS IKPNGRHLFT LDQNNGEVRV NGTLDYEENK FYKIEVQATD K Highest antigen sequence identity to the following orthologs - mouse 79%, rat 81%.

Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHA11
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCDHA11
Gene Accession No. Q9Y5I1
Gene Alias A830022B16Rik; cadherin-related neuronal receptor 11; cadherin-related neuronal receptor 7; CNR7; CNRN7; CNRS7; CRNR7; KIAA0345-like 3; LOW QUALITY PROTEIN: protocadherin alpha-10; PCDHA11; PCDH-ALPHA11; PCDH-alpha-11; PCDH-alpha9 homolog; protocadherin alpha 11; protocadherin alpha 9 homolog; protocadherin alpha-11
Gene Symbols PCDHA11
Host Species Rabbit
Immunogen Recombinant Human PCDHA11. Recombinant protein control fragment (Product #RP-108107).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56138
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.