Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDHA12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583290
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583290 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583290 Supplier Invitrogen™ Supplier No. PA583290
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNGEISYGIK MILPVSEKCM FSINPDTGEI RIYGELDFEE NNAYEIQVNA IDKGIPSMAG HSMVLVEV Highest antigen sequence indentity to the following orthologs: Mouse - 49%, Rat - 50%.

This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. The tandem array of 15 N-terminal exons, or variable exons, are followed by downstream C-terminal exons, or constant exons, which are shared by all genes in the cluster. The large, uninterrupted N-terminal exons each encode six cadherin ectodomains while the C-terminal exons encode the cytoplasmic domain. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins that most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been observed and additional variants have been suggested but their full-length nature has yet to be determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHA12
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCDHA12
Gene Accession No. Q9UN75
Gene Alias cadherin-related neuronal receptor 5; Cnr5; Crnr5; KIAA0345-like 2; MGC138485; MGC141932; PCDHA12; Pcdha13; PCDH-ALPHA12; PCDH-alpha-12; protocadherin alpha 12; protocadherin alpha 13; protocadherin alpha-12; rCNRv12
Gene Symbols PCDHA12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PCDHA12. Recombinant protein control fragment (Product #RP-103949).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56137
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.