Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCDHA13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577228
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.
Specifications
PCDHA13 | |
Polyclonal | |
Unconjugated | |
Pcdha13 | |
Cnr5; CNRN5; CNRS5; CRNR5; KIAA0345-like 1; ortholog of mouse CNR5; PCDHA13; PCDH-ALPHA13; PCDH-alpha-13; protocadherin alpha 13; protocadherin alpha-13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116742, 56136 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y5I0 | |
Pcdha13 | |
Recombinant full length Human PCDHA13. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction