Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDHA2 Monoclonal Antibody (2G12)
GREENER_CHOICE

Catalog No. PIMA522265
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522265 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522265 Supplier Invitrogen™ Supplier No. MA522265
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: SESLSLVLGK SLDREETAEV NLLLVATDGG KPELTGTVQI LIKVLDVNDN EPTFAQSVYK VKLLENTANG TLVVKLNASD ADEGPNSEIV YSLGSDVSST.

This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. The tandem array of 15 N-terminal exons, or variable exons, are followed by downstream C-terminal exons, or constant exons, which are shared by all genes in the cluster. The large, uninterrupted N-terminal exons each encode six cadherin ectodomains while the C-terminal exons encode the cytoplasmic domain. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins that most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been observed and additional variants have been suggested but their full-length nature has yet to be determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHA2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PCDHA2
Gene Accession No. Q9Y5H9
Gene Alias KIAA0345-like 12; PCDHA2; PCDH-ALPHA2; PCDH-alpha-2; protocadherin alpha 2; protocadherin alpha-2
Gene Symbols PCDHA2
Host Species Mouse
Immunogen PCDHA2 (NP_061728, 189 a.a. ~ 288 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56146
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.