Learn More
Invitrogen™ PCDHA4 Polyclonal Antibody

Description
Immunogen sequence: NEYFSLEKPPD DELVKGLGLI LRKSL Highest antigen sequence identity to the following orthologs - mouse 81%, rat 81%.
Specifications
Specifications
| Antigen | PCDHA4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PCDHA4 |
| Gene Accession No. | Q9UN74 |
| Gene Alias | cadherin-related neuronal receptor 1; cadherin-related neuronal receptor 4; Cnr1; CNRN1; Cnrv4; Crnr1; KIAA0345-like 10; MGC138307; MGC142169; ortholog of mouse CNR1; Pcdha4; PCDH-ALPHA4; PCDH-alpha-4; protocadherin alpha 4; protocadherin alpha-4; R75250 |
| Gene Symbols | PCDHA4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.