Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCDHA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542959
Description
Peptide sequence: SGYNAWLSYE LQPETASASI PFRVGLYTGE ISTTRALDET DAPRQRLLVL Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rat: 93%.
The gene encoding PCDHA4 is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. PCDHA4 is a single-pass type I membrane protein. It contains 6 cadherin domains. PCDHA4 is a potential calcium-dependent cell-adhesion protein. It may be involved in the establishment and maintenance of specific neuronal connections in the brain.
Specifications
| PCDHA4 | |
| Polyclonal | |
| Unconjugated | |
| PCDHA4 | |
| cadherin-related neuronal receptor 1; cadherin-related neuronal receptor 4; Cnr1; CNRN1; Cnrv4; Crnr1; KIAA0345-like 10; MGC138307; MGC142169; ortholog of mouse CNR1; Pcdha4; PCDH-ALPHA4; PCDH-alpha-4; protocadherin alpha 4; protocadherin alpha-4; R75250 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56144 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UN74 | |
| PCDHA4 | |
| Synthetic peptide directed towards the C-terminal of human PCDHA4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction