Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDHA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560748
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560748 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560748 Supplier Invitrogen™ Supplier No. PA560748
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GEIKVNGELD YEDYNSYEIN IDAMDKSTFP LSGHCKVVVK LLDVNDNTP Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 65%.

The gene encoding PCDHA5 is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. PCDHA5 is a single-pass type I membrane protein. It contains 6 cadherin domains. PCDHA5 is a potential calcium-dependent cell-adhesion protein. It may be involved in the establishment and maintenance of specific neuronal connections in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHA5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCDHA5
Gene Accession No. Q9Y5H7
Gene Alias cadherin-related neuronal receptor 5; cadherin-related neuronal receptor 6; Cnr6; CNRN6; CNRS6; Crnr6; KIAA0345-like 9; ortholog of mouse CNR6; PCDHA5; PCDH-ALPHA5; PCDH-alpha-5; protocadherin alpha 5; protocadherin alpha-5; rCNRv05
Gene Symbols PCDHA5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PCDHA5. Recombinant protein control fragment (Product #RP-96995).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56143
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.