Learn More
Invitrogen™ PCDHA6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110079
Description
Immunogen sequence: MVFTPEDRLG KQCLLLPLLL LAAWKVGSGQ LHYSVPEEAK HGTFVGRIAQ DLGLELAELV PRLFRMASKD REDLLEVNLQ NGILFVNSRI DREELCGRSA ECSIHLEVIV DRPLQVFHVD VEVRDINDNP PLFPVEEQRV LIYESRLPDS VFPLEGASDA DVGSNSILTY KLSSSEYFGL DVKINSDDNK QIGLLLKKSL DREEAPAHNL FLTATDGGKP ELTGTVQLLV.
This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. The tandem array of 15 N-terminal exons, or variable exons, are followed by downstream C-terminal exons, or constant exons, which are shared by all genes in the cluster. The large, uninterrupted N-terminal exons each encode six cadherin ectodomains while the C-terminal exons encode the cytoplasmic domain. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins that most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been observed and additional variants have been suggested but their full-length nature has yet to be determined.
Specifications
| PCDHA6 | |
| Polyclonal | |
| Unconjugated | |
| PCDHA6 | |
| cadherin-related neuronal receptor 2; cadherin-related neuronal receptor 6; CNR; CNR2; CNRN2; CNRS2; CRNR2; KIAA0345-like 8; PCDHA6; PCDH-ALPHA6; PCDH-alpha-6; protocadherin alpha 6; protocadherin alpha-6; rCNRv06 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12937, 393088, 56142 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.94 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UN73 | |
| PCDHA6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human PCDHA6 (NP_061732.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.