Learn More
Invitrogen™ PCDHA7 Monoclonal Antibody (1C7)

Description
Peptide Sequence: AESRPLDSRF PLEGASDADI GENALLTYRL SPNEYFFLDV PTSNQQVKPL GLVLRKLLDR EETPELHLLL TATDGGKPEL TGTVQLLITV LDNNDNAPV.
Specifications
Specifications
| Antigen | PCDHA7 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1C7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | PCDHA7 |
| Gene Accession No. | Q9UN72 |
| Gene Alias | CNR4; CNRN4; CNRS4; CRNR4; KIAA0345-like 7; ortholog to mouse CNR4; PCDHA7; PCDH-ALPHA7; PCDH-alpha-7; protocadherin alpha 7; protocadherin alpha-7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.