Learn More
Invitrogen™ PCDHA9 Monoclonal Antibody (3C1)

Description
Peptide Sequence: DVSPDIKSKF HMDPLSGAIT VIGHMDFEES RAHKIPVEAV DKGFPPLAGH CTLLVEVVDV NDNAPQLTIK TLSVPVKEDA QLGTVIALIS VIDLDADA.
Specifications
Specifications
| Antigen | PCDHA9 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3C1 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Pcdha9 |
| Gene Accession No. | Q9Y5H5 |
| Gene Alias | cadherin-related neuronal receptor 9; KIAA0345; KIAA0345-like 5; Pcdha9; PCDH-ALPHA9; PCDH-alpha-9; protocadherin alpha 9; protocadherin alpha-9; rCNRv09 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.