Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDHA9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548338
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548338 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548338 Supplier Invitrogen™ Supplier No. PA548338
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PCDHA9 Polyclonal Antibody for Western Blot, Flow

Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHA9
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Pcdha9
Gene Accession No. Q9Y5H5
Gene Alias cadherin-related neuronal receptor 9; KIAA0345; KIAA0345-like 5; Pcdha9; PCDH-ALPHA9; PCDH-alpha-9; protocadherin alpha 9; protocadherin alpha-9; rCNRv09
Gene Symbols Pcdha9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 226-253 amino acids from the N-terminal region of human PCDHA9.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9752
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.