Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDHAC1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen PCDHAC1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159214
SDP
View Documents
Novus Biologicals
NBP159214
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

PCDHAC1 Polyclonal specifically detects PCDHAC1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

PCDHAC1
Polyclonal
Rabbit
B2RNA7
56135
Synthetic peptides corresponding to PCDHAC1(protocadherin alpha subfamily C, 1) The peptide sequence was selected from the N terminal of PCDHAC1. Peptide sequence RVQALDPDEGSNGEVQYSLSNSTQAELRHRFHVHPKSGEVQVAASLGPPE.
Primary
Western Blot
Unconjugated
RUO
PCDH-ALPHA-C1, protocadherin alpha subfamily C, 1, protocadherin alpha-C1
PCDHAC1
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.