Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCDHAC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550723
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550723 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550723 Supplier Invitrogen™ Supplier No. PA550723
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total PCDHAC1 protein.

This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHAC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PCDHAC1
Gene Accession No. Q9H158
Gene Alias cadherin-related neuronal receptor c1; CNRc1; PCDHAC1; PCDH-ALPHA-C1; protocadherin alpha C1; protocadherin alpha subfamily C, 1; protocadherin alpha-C1; rCNRvc1
Gene Symbols PCDHAC1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human protocadherin alpha subfamily C, 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56135
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.