Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDHAC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565739
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565739 Supplier Thermo Scientific Supplier No. PA565739
Add to Cart
Add to Cart

PCDHAC1 Polyclonal Antibody

PCDHAC1 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHAC1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCDHAC1
Gene Accession No. Q9H158
Gene Alias cadherin-related neuronal receptor c1; CNRc1; PCDHAC1; PCDH-ALPHA-C1; protocadherin alpha C1; protocadherin alpha subfamily C, 1; protocadherin alpha-C1; rCNRvc1
Gene Symbols PCDHAC1
Host Species Rabbit
Immunogen Recombinant Human PCDHAC1. Recombinant protein control fragment (Product #RP-106415).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56135
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.